siRNA Details
Gene Symbol: Gm15506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052909.1 | - | 2 | 282 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB662268.1 | - | 2 | 282 | Chr. 7 - 98682840 - 98702928 [-] on Build GRCm38 |
