Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MIR100HG SIRNA Assays ›
          Gene Symbol:
          MIR100HG
          Gene Name:
          mir-100-let-7a-2 cluster host gene
          Gene Aliases:
          AGD1, linc-NeD125, lncRNA-N2
          Chromosome Location:
          Chr.11: 122089103 - 122203062 on Build GRCh38
          UniGene ID:
          Hs.44098
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n542638

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MIR100HG
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024430.2 4 1016
          NR_137175.1 2 1287
          NR_137176.1 2 998
          NR_137177.1 2 977
          NR_137179.1 5 970
          NR_137180.1 5 1028
          NR_137181.1 3 913
          NR_137182.1 4 995
          NR_137183.1 3 904
          NR_137184.1 2 822
          NR_137185.1 3 883
          NR_137186.1 3 792
          NR_137187.1 3 771
          NR_137188.1 3 861
          NR_137189.1 2 1308
          NR_137190.1 3 1341
          NR_137191.1 3 1320
          NR_137192.1 4 1180
          NR_137193.1 5 1007
          NR_137194.1 3 882
          NR_137195.1 2 1050
          NR_137196.1 2 1071
          GenBank mRNA AB075498.1 - 2 1540
          AB485963.1 - 4 899
          AK021552.1 - 1 182
          AK091713.1 - 3 672
          AK123947.1 - 2 1224
          AL832469.1 - 2 604
          BC040599.1 - 2 600
          BX538058.1 - 2 577
          BX647608.1 - 2 578
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AB485964.1 - 3 751 Chr. 11 - 122091162 - 122157093 [-] on Build GRCh38
          GenBankmRNA AB485961.1 - 2 765 Chr. 11 - 122091162 - 122101686 [-] on Build GRCh38
          GenBankmRNA AB485960.1 - 2 609 Chr. 11 - 122091162 - 122100478 [-] on Build GRCh38
          GenBankmRNA DA778681.1 - 2 569 Chr. 11 - 122091204 - 122100438 [-] on Build GRCh38
          GenBankmRNA AB485962.1 - 3 656 Chr. 11 - 122091162 - 122180403 [-] on Build GRCh38
          GenBankmRNA DA769648.1 - 3 499 Chr. 11 - 122091136 - 122101686 [-] on Build GRCh38
          GenBankmRNA DA787193.1 - 2 591 Chr. 11 - 122091205 - 122100439 [-] on Build GRCh38
          GenBankmRNA DA573353.1 - 2 590 Chr. 11 - 122090936 - 122100438 [-] on Build GRCh38
          GenBankmRNA BE883292.1 - 2 611 Chr. 11 - 122091092 - 122100480 [-] on Build GRCh38
          GenBankmRNA DA783606.1 - 2 571 Chr. 11 - 122091200 - 122100436 [-] on Build GRCh38
          GenBankmRNA BI463061.1 - 2 605 Chr. 11 - 122091189 - 122100448 [-] on Build GRCh38
          GenBankmRNA DA483192.1 - 2 569 Chr. 11 - 122090961 - 122100438 [-] on Build GRCh38
          GenBankmRNA BJ997524.1 - 2 579 Chr. 11 - 122091195 - 122100448 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-cps46:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0