Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SVIL-AS1 SIRNA Assays ›
          Gene Symbol:
          SVIL-AS1
          Gene Name:
          SVIL antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 29409534 - 29487856 on Build GRCh38
          UniGene ID:
          Hs.499209
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n544030

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SVIL-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003930.2 1 100
          NR_110920.1 1 100
          NR_110921.1 1 100
          NR_110922.1 1 100
          NR_110923.1 1 100
          NR_110924.1 1 100
          NR_110925.1 1 100
          NR_110926.1 1 100
          NR_110927.1 1 100
          NR_110929.1 1 100
          GenBank mRNA BC031656.1 - 1 55
          BC066966.1 - 1 98
          BC107795.1 - 1 84
          BM925434.1 - 1 67
          DA024320.1 - 1 100
          DA715011.1 - 1 77
          DA745252.1 - 1 45
          DA952579.1 - 1 98
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AV710390.1 - 1 76 Chr. 10 - 29409558 - 29483504 [+] on Build GRCh38
          GenBankmRNA DA802966.1 - 1 77 Chr. 10 - 29409557 - 29510818 [+] on Build GRCh38
          GenBankmRNA DA121252.1 - 1 76 Chr. 10 - 29409558 - 29483221 [+] on Build GRCh38
          GenBankmRNA DA661027.1 - 1 98 Chr. 10 - 29409536 - 29420878 [+] on Build GRCh38
          GenBankmRNA DA581920.1 - 1 77 Chr. 10 - 29409557 - 29420885 [+] on Build GRCh38
          GenBankmRNA HY001946.1 - 1 78 Chr. 10 - 29409556 - 29454522 [+] on Build GRCh38
          GenBankmRNA DA177235.1 - 1 45 Chr. 10 - 29409589 - 29420684 [+] on Build GRCh38
          GenBankmRNA DB180401.1 - 1 96 Chr. 10 - 29409538 - 29420869 [+] on Build GRCh38
          GenBankmRNA W56452.1 - 1 233 Chr. 10 - 29409402 - 29458400 [+] on Build GRCh38
          GenBankmRNA DB262217.1 - 1 77 Chr. 10 - 29409557 - 29420897 [+] on Build GRCh38
          GenBankmRNA DA322935.1 - 1 100 Chr. 10 - 29409534 - 29420775 [+] on Build GRCh38
          GenBankmRNA DB047449.1 - 1 70 Chr. 10 - 29409564 - 29420900 [+] on Build GRCh38
          GenBankmRNA CB960614.1 - 1 104 Chr. 10 - 29409557 - 29415958 [+] on Build GRCh38
          GenBankmRNA DA507912.1 - 1 77 Chr. 10 - 29409557 - 29483252 [+] on Build GRCh38
          GenBankmRNA DA888030.1 - 1 77 Chr. 10 - 29409557 - 29420895 [+] on Build GRCh38
          GenBankmRNA DA265970.1 - 1 71 Chr. 10 - 29409564 - 29487550 [+] on Build GRCh38
          GenBankmRNA DB099389.1 - 1 100 Chr. 10 - 29409534 - 29420897 [+] on Build GRCh38
          GenBankmRNA DB049688.1 - 1 70 Chr. 10 - 29409564 - 29420901 [+] on Build GRCh38
          GenBankmRNA DB031339.1 - 1 98 Chr. 10 - 29409537 - 29420901 [+] on Build GRCh38
          GenBankmRNA DA307490.1 - 1 100 Chr. 10 - 29409534 - 29483247 [+] on Build GRCh38
          GenBankmRNA DA110605.1 - 1 100 Chr. 10 - 29409534 - 29470500 [+] on Build GRCh38
          GenBankmRNA BG036994.1 - 1 103 Chr. 10 - 29409539 - 29487715 [+] on Build GRCh38
          GenBankmRNA DA373323.1 - 1 77 Chr. 10 - 29409557 - 29420883 [+] on Build GRCh38
          GenBankmRNA DA591198.1 - 1 100 Chr. 10 - 29409534 - 29420881 [+] on Build GRCh38
          GenBankmRNA DA580038.1 - 1 70 Chr. 10 - 29409564 - 29420906 [+] on Build GRCh38
          GenBankmRNA DB284993.1 - 1 77 Chr. 10 - 29409557 - 29420877 [+] on Build GRCh38
          GenBankmRNA DA119810.1 - 1 87 Chr. 10 - 29409547 - 29458376 [+] on Build GRCh38
          GenBankmRNA BM565031.1 - 1 44 Chr. 10 - 29409597 - 29420946 [+] on Build GRCh38
          GenBankmRNA DB012592.1 - 1 73 Chr. 10 - 29409561 - 29420899 [+] on Build GRCh38
          GenBankmRNA BI756656.1 - 1 62 Chr. 10 - 29409572 - 29420950 [+] on Build GRCh38
          GenBankmRNA DA168383.1 - 1 100 Chr. 10 - 29409534 - 29420880 [+] on Build GRCh38
          GenBankmRNA CB988224.1 - 1 104 Chr. 10 - 29409557 - 29421050 [+] on Build GRCh38
          GenBankmRNA BM511177.1 - 1 44 Chr. 10 - 29409597 - 29420778 [+] on Build GRCh38
          GenBankmRNA DA449336.1 - 1 100 Chr. 10 - 29409534 - 29420863 [+] on Build GRCh38
          GenBankmRNA CB046493.1 - 1 36 Chr. 10 - 29409607 - 29458607 [+] on Build GRCh38
          GenBankmRNA DB003348.1 - 1 100 Chr. 10 - 29409534 - 29420847 [+] on Build GRCh38
          GenBankmRNA DA836156.1 - 1 35 Chr. 10 - 29409599 - 29420931 [+] on Build GRCh38
          GenBankmRNA DB167786.1 - 1 45 Chr. 10 - 29409589 - 29483291 [+] on Build GRCh38
          GenBankmRNA BM835655.1 - 1 57 Chr. 10 - 29409577 - 29420879 [+] on Build GRCh38
          GenBankmRNA DA177718.1 - 1 97 Chr. 10 - 29409538 - 29487538 [+] on Build GRCh38
          GenBankmRNA CK823803.1 - 1 44 Chr. 10 - 29409597 - 29420937 [+] on Build GRCh38
          GenBankmRNA DA570840.1 - 558 Chr. - - [-] on Build
          GenBankmRNA DA536274.1 - 1 45 Chr. 10 - 29409589 - 29415881 [+] on Build GRCh38
          GenBankmRNA DA548063.1 - 1 100 Chr. 10 - 29409534 - 29420868 [+] on Build GRCh38
          GenBankmRNA DB042440.1 - 1 70 Chr. 10 - 29409564 - 29420876 [+] on Build GRCh38
          GenBankmRNA DA405633.1 - 1 96 Chr. 10 - 29409538 - 29420863 [+] on Build GRCh38
          GenBankmRNA HY158995.1 - 1 75 Chr. 10 - 29409559 - 29458600 [+] on Build GRCh38
          GenBankmRNA DA831992.1 - 1 71 Chr. 10 - 29409564 - 29421166 [+] on Build GRCh38
          GenBankmRNA DB091461.1 - 1 96 Chr. 10 - 29409538 - 29420869 [+] on Build GRCh38
          GenBankmRNA BI820719.1 - 1 54 Chr. 10 - 29409580 - 29421127 [+] on Build GRCh38
          GenBankmRNA DA003665.1 - 1 70 Chr. 10 - 29409564 - 29420919 [+] on Build GRCh38
          GenBankmRNA BI916268.1 - 1 43 Chr. 10 - 29409591 - 29483451 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0