Search
Search

ERVK13-1| Gene Symbol: ERVK13-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040023.1 | 3 | 5074 | |||
| GenBank mRNA | AK000400.1 | - | 1 | 744 | ||
| AK091794.1 | - | 3 | 2866 | |||
| AK095440.1 | - | 1 | 2671 | |||
| AK097909.1 | - | 1 | 1129 | |||
| AK131509.1 | - | 1 | 3700 | |||
| AK292678.1 | - | 1 | 1647 | |||
| AX747190.1 | - | 3 | 2866 | |||
| BC043252.1 | - | 3 | 5076 | |||