siRNA Details
Gene Symbol: LOC101928323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125400.1 | 1 | 114 | |||
GenBank mRNA | HY122338.1 | - | 1 | 115 |

Gene Symbol: LOC101928323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125400.1 | 1 | 114 | |||
GenBank mRNA | HY122338.1 | - | 1 | 115 |