Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG3 SIRNA Assays ›
          Gene Symbol:
          SNHG3 RCC1
          Gene Name:
          small nucleolar RNA host gene 3
          regulator of chromosome condensation 1
          Gene Aliases:
          NCRNA00014, RNU17C, RNU17D, U17HG, U17HG-A, U17HG-AB
          CHC1, RCC1-I, SNHG3-RCC1
          Chromosome Location:
          Chr.1: 28505943 - 28510892 on Build GRCh38
          Chromosome Location:
          Chr.1: 28505943 - 28539196 on Build GRCh38
          UniGene ID:
          Hs.469723
          Hs.469723
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n547163

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNHG3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002909.2
          3 250
          NR_036473.1
          4 358
          GenBank mRNA AJ006834.1 - LIT1316
          U17HG
          3 150
          AJ006835.1 - LIT1315
          U17HG
          3 150
          AK092726.1 -
          2 1883
          BC006790.1 -
          3 132
          BC012122.1 -
          3 138
          BC017334.2 -
          3 125
          DA044266.1 -
          3 209
          DA135739.1 -
          4 321
          Gene Symbol: RCC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BF667678.1 - 3 248
          CB985055.1 - 3 216
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM479709.1 - 3 135 Chr. 1 - 28506059 - 28509634 [+] on Build GRCh38
          GenBankmRNA DA820174.1 - 3 147 Chr. 1 - 28506046 - 28509321 [+] on Build GRCh38
          GenBankmRNA CN366536.1 - 2 105 Chr. 1 - 28508133 - 28509423 [+] on Build GRCh38
          GenBankmRNA HY173512.1 - 3 149 Chr. 1 - 28506045 - 28509232 [+] on Build GRCh38
          GenBankmRNA HY075800.1 - 3 151 Chr. 1 - 28506045 - 28509246 [+] on Build GRCh38
          GenBankmRNA AA352342.1 - 2 100 Chr. 1 - 28508136 - 28509153 [+] on Build GRCh38
          GenBankmRNA HY180673.1 - 3 148 Chr. 1 - 28506045 - 28509192 [+] on Build GRCh38
          GenBankmRNA BI833284.1 - 3 131 Chr. 1 - 28506060 - 28509531 [+] on Build GRCh38
          GenBankmRNA BM788818.1 - 2 126 Chr. 1 - 28508127 - 28509339 [+] on Build GRCh38
          GenBankmRNA BU183513.1 - 3 137 Chr. 1 - 28506056 - 28509509 [+] on Build GRCh38
          GenBankmRNA BM450342.1 - 127 Chr. - - [-] on Build
          GenBankmRNA HY113224.1 - 3 206 Chr. 1 - 28505989 - 28509206 [+] on Build GRCh38
          GenBankmRNA DA998284.1 - 3 156 Chr. 1 - 28506045 - 28509318 [+] on Build GRCh38
          GenBankmRNA AL523116.3 - 2 121 Chr. 1 - 28508127 - 28509940 [+] on Build GRCh38
          GenBankmRNA CN366544.1 - 2 119 Chr. 1 - 28508127 - 28509283 [+] on Build GRCh38
          GenBankmRNA BM834932.1 - 1 29 Chr. 1 - 28508876 - 28509340 [+] on Build GRCh38
          GenBankmRNA BU943197.1 - 3 152 Chr. 1 - 28506044 - 28509509 [+] on Build GRCh38
          GenBankmRNA CN366542.1 - 3 138 Chr. 1 - 28506055 - 28509581 [+] on Build GRCh38
          GenBankmRNA HY153255.1 - 3 148 Chr. 1 - 28506045 - 28509198 [+] on Build GRCh38
          GenBankmRNA DA933716.1 - 1 84 Chr. 1 - 28508824 - 28509359 [+] on Build GRCh38
          GenBankmRNA CN366540.1 - 2 135 Chr. 1 - 28508127 - 28509421 [+] on Build GRCh38
          GenBankmRNA HY084156.1 - 3 148 Chr. 1 - 28506045 - 28509217 [+] on Build GRCh38
          GenBankmRNA DB128729.1 - 3 149 Chr. 1 - 28506044 - 28509326 [+] on Build GRCh38
          GenBankmRNA BF941702.1 - 3 143 Chr. 1 - 28506050 - 28509152 [+] on Build GRCh38
          GenBankmRNA CN366538.1 - 3 150 Chr. 1 - 28506055 - 28509488 [+] on Build GRCh38
          GenBankmRNA BG474710.1 - 3 139 Chr. 1 - 28506055 - 28509618 [+] on Build GRCh38
          GenBankmRNA HY182836.1 - 3 148 Chr. 1 - 28506045 - 28509242 [+] on Build GRCh38
          GenBankmRNA CB157773.1 - 3 138 Chr. 1 - 28506055 - 28509221 [+] on Build GRCh38
          GenBankmRNA AA352343.1 - 2 100 Chr. 1 - 28508136 - 28509163 [+] on Build GRCh38
          GenBankmRNA BF795420.1 - 137 Chr. - - [-] on Build
          GenBankmRNA HY112100.1 - 3 225 Chr. 1 - 28505969 - 28509173 [+] on Build GRCh38
          GenBankmRNA CB985655.1 - 3 153 Chr. 1 - 28506043 - 28509509 [+] on Build GRCh38
          GenBankmRNA HY059835.1 - 3 148 Chr. 1 - 28506045 - 28509325 [+] on Build GRCh38
          GenBankmRNA HY089919.1 - 3 148 Chr. 1 - 28506045 - 28509246 [+] on Build GRCh38
          GenBankmRNA CB963044.1 - 3 204 Chr. 1 - 28505990 - 28509462 [+] on Build GRCh38
          GenBankmRNA HY179907.1 - 3 148 Chr. 1 - 28506045 - 28509261 [+] on Build GRCh38
          GenBankmRNA DA968918.1 - 3 149 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA BM759606.1 - 3 150 Chr. 1 - 28506043 - 28509424 [+] on Build GRCh38
          GenBankmRNA BM802102.1 - 137 Chr. - - [-] on Build
          GenBankmRNA BF982554.1 - 2 98 Chr. 1 - 28508137 - 28509570 [+] on Build GRCh38
          GenBankmRNA DA787816.1 - 3 149 Chr. 1 - 28506044 - 28509270 [+] on Build GRCh38
          GenBankmRNA CN366535.1 - 3 140 Chr. 1 - 28506053 - 28509489 [+] on Build GRCh38
          GenBankmRNA BG023899.1 - 119 Chr. - - [-] on Build
          GenBankmRNA BM790281.1 - 3 137 Chr. 1 - 28506056 - 28509323 [+] on Build GRCh38
          GenBankmRNA AV763089.1 - 2 143 Chr. 1 - 28508127 - 28509528 [+] on Build GRCh38
          GenBankmRNA CD048541.1 - 3 169 Chr. 1 - 28506045 - 28509614 [+] on Build GRCh38
          GenBankmRNA HY176989.1 - 3 148 Chr. 1 - 28506045 - 28509229 [+] on Build GRCh38
          GenBankmRNA DA478694.1 - 3 149 Chr. 1 - 28506044 - 28509328 [+] on Build GRCh38
          GenBankmRNA HY175139.1 - 3 148 Chr. 1 - 28506045 - 28509219 [+] on Build GRCh38
          GenBankmRNA CN366543.1 - 3 146 Chr. 1 - 28506055 - 28509366 [+] on Build GRCh38
          GenBankmRNA DA089660.1 - 3 209 Chr. 1 - 28505984 - 28509274 [+] on Build GRCh38
          GenBankmRNA HY176129.1 - 3 148 Chr. 1 - 28506045 - 28509283 [+] on Build GRCh38
          GenBankmRNA BY796950.2 - 2 118 Chr. 1 - 28508127 - 28509414 [+] on Build GRCh38
          GenBankmRNA DA704998.1 - 3 149 Chr. 1 - 28506044 - 28509252 [+] on Build GRCh38
          GenBankmRNA CN366541.1 - 2 106 Chr. 1 - 28508130 - 28509185 [+] on Build GRCh38
          GenBankmRNA AV689452.1 - 3 184 Chr. 1 - 28506009 - 28509366 [+] on Build GRCh38
          GenBankmRNA DA728986.1 - 3 149 Chr. 1 - 28506044 - 28509467 [+] on Build GRCh38
          GenBankmRNA DA381375.1 - 1 193 Chr. 1 - 28508764 - 28509340 [+] on Build GRCh38
          GenBankmRNA BQ224438.1 - 3 149 Chr. 1 - 28506055 - 28509617 [+] on Build GRCh38
          GenBankmRNA BE867363.1 - 2 123 Chr. 1 - 28508131 - 28509511 [+] on Build GRCh38
          GenBankmRNA CN366539.1 - 3 125 Chr. 1 - 28506068 - 28509242 [+] on Build GRCh38
          GenBankmRNA HY182945.1 - 3 148 Chr. 1 - 28506045 - 28509230 [+] on Build GRCh38
          GenBankmRNA BG115368.1 - 2 139 Chr. 1 - 28508127 - 28509208 [+] on Build GRCh38
          GenBankmRNA BG028447.1 - 3 135 Chr. 1 - 28506057 - 28509522 [+] on Build GRCh38
          GenBankmRNA CB128113.1 - 3 152 Chr. 1 - 28506045 - 28509219 [+] on Build GRCh38
          GenBankmRNA HY177570.1 - 3 148 Chr. 1 - 28506045 - 28509225 [+] on Build GRCh38
          GenBankmRNA BG291633.1 - 3 130 Chr. 1 - 28506067 - 28509489 [+] on Build GRCh38
          GenBankmRNA BM771983.1 - 3 149 Chr. 1 - 28506044 - 28509374 [+] on Build GRCh38
          GenBankmRNA BU187480.1 - 2 122 Chr. 1 - 28508127 - 28509634 [+] on Build GRCh38
          GenBankmRNA CB163464.1 - 3 133 Chr. 1 - 28506060 - 28509328 [+] on Build GRCh38
          GenBankmRNA BI833122.1 - 3 132 Chr. 1 - 28506059 - 28509542 [+] on Build GRCh38
          GenBankmRNA BI772556.1 - 2 133 Chr. 1 - 28508116 - 28509484 [+] on Build GRCh38
          GenBankmRNA DA056610.1 - 3 209 Chr. 1 - 28505984 - 28509252 [+] on Build GRCh38
          GenBankmRNA BU198833.1 - 3 153 Chr. 1 - 28506044 - 28509422 [+] on Build GRCh38
          GenBankmRNA BM802666.1 - 137 Chr. - - [-] on Build
          GenBankmRNA HY179875.1 - 3 148 Chr. 1 - 28506045 - 28509263 [+] on Build GRCh38
          GenBankmRNA AV764560.2 - 3 142 Chr. 1 - 28506051 - 28509412 [+] on Build GRCh38
          GenBankmRNA BF036396.1 - 2 115 Chr. 1 - 28508127 - 28509376 [+] on Build GRCh38
          GenBankmRNA BQ218931.1 - 3 133 Chr. 1 - 28506060 - 28509544 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0