Search
Search

DAB1-AS1| Gene Symbol: DAB1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104362.1 | 2 | 794 | |||
| NR_104363.1 | 2 | 680 | ||||
| NR_104364.1 | 2 | 670 | ||||
| NR_104365.1 | 2 | 633 | ||||
| GenBank mRNA | BC033978.1 | - | 2 | 588 | ||
| BC047487.1 | - | 2 | 794 | |||