Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HAGLR SIRNA Assays ›
          Gene Symbol:
          HAGLR
          Gene Name:
          HOXD antisense growth-associated long non-coding RNA
          Gene Aliases:
          HOXD-AS1, Mdgt
          Chromosome Location:
          Chr.2: 176173189 - 176188958 on Build GRCh38
          UniGene ID:
          Hs.98661
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n549838

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HAGLR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_033979.2 3 405
          NR_110458.1 3 679
          NR_110459.1 5 630
          NR_110460.1 5 600
          NR_110461.1 4 535
          NR_110462.1 2 516
          NR_110463.1 4 498
          NR_110464.1 4 486
          NR_110465.1 4 484
          NR_110466.1 3 414
          NR_110467.1 3 387
          NR_110468.1 3 375
          GenBank mRNA BC030713.2 - 3 666
          BC047481.1 - 3 666
          BG721643.1 - 3 674
          CB153479.1 - 3 405
          DA326484.1 - 5 590
          DA398428.1 - 4 391
          DA736032.1 - 4 536
          DB045673.1 - 3 342
          DB067012.1 - 3 371
          DB100342.1 - 4 452
          DB100521.1 - 4 399
          HY138745.1 - 3 416
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA401217.1 - 2 334 Chr. 2 - 176176476 - 176177568 [-] on Build GRCh38
          GenBankmRNA DA331336.1 - 3 374 Chr. 2 - 176176386 - 176188531 [-] on Build GRCh38
          GenBankmRNA DA727022.1 - 4 530 Chr. 2 - 176176553 - 176188264 [-] on Build GRCh38
          GenBankmRNA DA292668.1 - 3 350 Chr. 2 - 176176362 - 176188508 [-] on Build GRCh38
          GenBankmRNA BG718435.1 - 3 663 Chr. 2 - 176176481 - 176178109 [-] on Build GRCh38
          GenBankmRNA DA755456.1 - 3 405 Chr. 2 - 176176460 - 176188958 [-] on Build GRCh38
          GenBankmRNA BE378592.1 - 4 408 Chr. 2 - 176176487 - 176179060 [-] on Build GRCh38
          GenBankmRNA BG717191.1 - 2 519 Chr. 2 - 176176472 - 176178125 [-] on Build GRCh38
          GenBankmRNA BG324915.1 - 365 Chr. - - [-] on Build
          GenBankmRNA BI520150.1 - 3 669 Chr. 2 - 176176399 - 176178119 [-] on Build GRCh38
          GenBankmRNA HY025996.1 - 3 347 Chr. 2 - 176164051 - 176178099 [-] on Build GRCh38
          GenBankmRNA DA182774.1 - 3 368 Chr. 2 - 176176390 - 176188526 [-] on Build GRCh38
          GenBankmRNA AL120749.1 - 3 284 Chr. 2 - 176176169 - 176188442 [-] on Build GRCh38
          GenBankmRNA HY008350.1 - 3 364 Chr. 2 - 176176388 - 176178118 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nglt7:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0