Search
Search

2610305D13Rik| Gene Symbol: 2610305D13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145078.2 | NP_659544.2 | 5 | 1310 | ||
| GenBank mRNA | AK011986.1 | - | 2 | 1012 | ||
| BC032186.1 | - | 1 | 798 | |||
| BC058858.1 | - | 5 | 1314 | |||
| BC138179.1 | - | 2 | 953 | 8861198 | ||
| BC145854.1 | - | 2 | 953 | 40131157 | ||