Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aipl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053245.2 | NP_444475.2 | 2 | 199 | ||
GenBank mRNA | AF296412.1 | - | 2 | 148 | ||
BC028285.1 | - | 2 | 199 | 4501401 | ||
CK618559.1 | - | 2 | 199 |