Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PAK1 SIRNA Assays ›
          Gene Symbol:
          PAK1
          Gene Name:
          p21 (RAC1) activated kinase 1
          Gene Aliases:
          PAKalpha
          Chromosome Location:
          Chr.11: 77322015 - 77474063 on Build GRCh38
          UniGene ID:
          Hs.435714
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s10019
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PAK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001128620.1 NP_001122092.1 12, 13 1743
          NM_002576.4 NP_002567.3 12, 13 1743
          XM_011545080.2 XP_011543382.1 12, 13 1450
          XM_011545081.2 XP_011543383.1 12, 13 1519
          XM_011545082.2 XP_011543384.1 13, 14 1473
          XM_011545083.2 XP_011543385.1 12, 13 1740
          XM_011545084.2 XP_011543386.1 12, 13 1310
          XM_017017846.1 XP_016873335.1 12, 13 1252
          XM_017017847.1 XP_016873336.1 12, 13 1450
          XM_017017848.1 XP_016873337.1 12, 13 1519
          XM_017017849.1 XP_016873338.1 12, 13 1396
          XM_017017850.1 XP_016873339.1 13, 14 1459
          GenBank mRNA AF071884.1 - 11, 12 1221
          AK055228.1 - 12, 13 1410
          AK293098.1 - 12, 13 1399
          AK295344.1 - 5, 6 434
          AK296006.1 - 11, 12 1151
          BC050377.1 - 12, 13 1335
          BC109298.1 - 12, 13 1351
          BC109299.1 - 12, 13 1351 40034484
          U24152.1 - 12, 13 1603
          U51120.1 - 12, 13 1264

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00176815_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          mitotic cell cycle
          response to hypoxia
          positive regulation of protein phosphorylation
          stimulatory C-type lectin receptor signaling pathway
          protein phosphorylation
          exocytosis
          apoptotic process
          axonogenesis
          positive regulation of cell proliferation
          positive regulation of cell migration
          T cell costimulation
          actin cytoskeleton reorganization
          cellular response to insulin stimulus
          positive regulation of peptidyl-serine phosphorylation
          positive regulation of intracellular estrogen receptor signaling pathway
          Fc-epsilon receptor signaling pathway
          Fc-gamma receptor signaling pathway involved in phagocytosis
          wound healing
          positive regulation of JUN kinase activity
          protein autophosphorylation
          ephrin receptor signaling pathway
          branching morphogenesis of an epithelial tube
          neuron projection morphogenesis
          T cell receptor signaling pathway
          positive regulation of stress fiber assembly
          negative regulation of cell proliferation involved in contact inhibition
          protein kinase activity
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          protein binding
          collagen binding
          ATP binding
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0