Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAK3 SIRNA Assays ›
Gene Symbol:
PAK3
Gene Name:
p21 (RAC1) activated kinase 3
Gene Aliases:
ARA, MRX30, MRX47, OPHN3, PAK-3, PAK3beta, bPAK, beta-PAK
Chromosome Location:
Chr.X: 110944241 - 111227361 on Build GRCh38
UniGene ID:
Hs.593599
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s10026
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAK3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128166.2
NP_001121638.1
11
1406
NM_001128167.2
NP_001121639.1
10
1333
NM_001128168.2
NP_001121640.1
15
1655
NM_001128172.2
NP_001121644.1
10
1008
NM_001128173.2
NP_001121645.1
14
1592
NM_001324325.1
NP_001311254.1
11
1484
NM_001324326.1
NP_001311255.1
11
1329
NM_001324327.1
NP_001311256.1
13
1441
NM_001324328.1
NP_001311257.1
12
1374
NM_001324329.1
NP_001311258.1
14
1589
NM_001324330.1
NP_001311259.1
11
1481
NM_001324331.1
NP_001311260.1
12
1396
NM_001324332.1
NP_001311261.1
13
1547
NM_001324333.1
NP_001311262.1
13
1522
NM_001324334.1
NP_001311263.1
12
1477
NM_002578.4
NP_002569.1
13
1544
NR_136747.1
12
1396
NR_136748.1
10
1093
XM_005262132.2
XP_005262189.1
14
1584
XM_005262133.3
XP_005262190.1
13
1539
XM_006724654.2
XP_006724717.1
13
1431
XM_006724655.2
XP_006724718.1
12
1374
XM_011530962.1
XP_011529264.1
14
1582
XM_011530967.2
XP_011529269.1
13
1522
XM_011530968.2
XP_011529270.1
14
1598
XM_011530969.1
XP_011529271.1
12
1443
XM_017029557.1
XP_016885046.1
12
1443
XM_017029558.1
XP_016885047.1
12
1386
XM_017029559.1
XP_016885048.1
11
1398
XM_017029560.1
XP_016885049.1
11
1329
XM_017029561.1
XP_016885050.1
13
1546
XM_017029562.1
XP_016885051.1
14
1584
XM_017029563.1
XP_016885052.1
15
1647
GenBank mRNA
AB102659.1
-
10
963
AF068864.1
-
9
918
AF155651.1
-
13
1309
AK128022.1
-
12
1651
AK290504.1
-
10
1333
AK315284.1
-
11
1250
AM943850.1
-
10
963
AM943851.1
-
10
981
AM943852.1
-
11
1026
BC117353.1
-
10
1012
BC117355.1
-
10
1012
BC143544.1
-
10
1012
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00176828_m1
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
mitotic cell cycle
stimulatory C-type lectin receptor signaling pathway
axonogenesis
dendrite development
regulation of actin filament polymerization
T cell costimulation
positive regulation of neuron apoptotic process
ephrin receptor signaling pathway
synapse organization
T cell receptor signaling pathway
dendritic spine morphogenesis
positive regulation of dendritic spine morphogenesis
cellular response to organic cyclic compound
positive regulation of DNA biosynthetic process
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase activity
protein binding
ATP binding
Rho GTPase binding
SH3 domain binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0