Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttyh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053273.2 | NP_444503.2 | 13 | 1553 | ||
GenBank mRNA | AF329682.1 | - | 13 | 1493 | ||
AK152172.1 | - | 13 | 1505 | |||
AK154835.1 | - | 13 | 1560 | |||
AK155166.1 | - | 13 | 1554 | |||
AK156393.1 | - | 12 | 1320 | |||
AK168408.1 | - | 13 | 1506 | |||
AK184196.1 | - | 3 | 251 | |||
BC060740.1 | - | 13 | 1496 | 4158813 |