Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Card14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130886.3 | NP_570956.1 | 19 | 2833 | ||
GenBank mRNA | AF363457.1 | - | 19 | 2812 | ||
AK028888.1 | - | 13 | 2218 | |||
BC004692.1 | - | 15 | 2204 | |||
BC029102.1 | - | 19 | 2833 | 3979883 |