Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285920.1 | NP_001272849.1 | 12 | 1554 | ||
NM_001285921.1 | NP_001272850.1 | 12 | 1537 | |||
NM_001285922.1 | NP_001272851.1 | 11 | 1468 | |||
NM_001285923.1 | NP_001272852.1 | 11 | 1468 | |||
NM_133201.3 | NP_573464.2 | 11 | 1471 | |||
GenBank mRNA | AB048831.1 | - | 9 | 1072 | ||
AF384100.1 | - | 12 | 1351 | |||
AK042080.1 | - | 11 | 1252 | |||
AK049583.1 | - | 11 | 1261 | |||
AK052689.1 | - | 11 | 1252 | |||
AK122220.1 | - | 10 | 2446 | |||
AK146801.1 | - | 12 | 1314 | |||
AK147863.1 | - | 11 | 1267 | |||
AK169373.1 | - | 11 | 1256 | |||
AY028170.1 | - | 1232 | ||||
AY123975.1 | - | 11 | 1418 | |||
BC046503.1 | - | 11 | 1249 | 4191179 |