Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133246.5 | NP_573509.1 | 6 | 735 | ||
GenBank mRNA | AF321128.1 | - | 6 | 674 | ||
AK036464.1 | - | 6 | 737 | |||
AY184359.1 | - | 6 | 689 | |||
BC117013.1 | - | 6 | 651 | 40126332 | ||
BC117015.1 | - | 6 | 651 | 40126334 | ||
BC144853.1 | - | 6 | 651 | 9053401 |