Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCM1 SIRNA Assays ›
Gene Symbol:
PCM1
Gene Name:
pericentriolar material 1
Gene Aliases:
PTC4, RET/PCM-1
Chromosome Location:
Chr.8: 17922763 - 18029948 on Build GRCh38
UniGene ID:
Hs.491148
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s10127
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001315507.1
NP_001302436.1
28
4968
NM_001315508.1
NP_001302437.1
27
4806
NM_006197.3
NP_006188.3
28
4968
XM_006716336.3
XP_006716399.1
31
5407
XM_006716340.3
XP_006716403.1
31
5407
XM_006716349.3
XP_006716412.1
27
4879
XM_011544528.2
XP_011542830.1
31
5428
XM_011544529.2
XP_011542831.1
33
5734
XM_011544530.2
XP_011542832.1
32
5550
XM_011544532.2
XP_011542834.1
31
5425
XM_011544533.2
XP_011542835.1
31
5428
XM_011544534.2
XP_011542836.1
31
5428
XM_011544535.2
XP_011542837.1
31
5428
XM_011544540.2
XP_011542842.1
31
5356
XM_017013472.1
XP_016868961.1
32
5544
XM_017013473.1
XP_016868962.1
32
5540
XM_017013474.1
XP_016868963.1
29
5158
XM_017013475.1
XP_016868964.1
31
5294
XM_017013476.1
XP_016868965.1
28
4878
XM_017013477.1
XP_016868966.1
31
5483
XM_017013478.1
XP_016868967.1
31
5461
XM_017013479.1
XP_016868968.1
31
5426
XM_017013480.1
XP_016868969.1
29
5098
XM_017013481.1
XP_016868970.1
31
5401
XM_017013482.1
XP_016868971.1
28
5062
XM_017013483.1
XP_016868972.1
30
5343
XM_017013484.1
XP_016868973.1
27
4769
XM_017013485.1
XP_016868974.1
30
5347
XM_017013486.1
XP_016868975.1
28
4993
XM_017013487.1
XP_016868976.1
28
4996
XM_017013488.1
XP_016868977.1
28
4993
XM_017013489.1
XP_016868978.1
30
5296
XM_017013490.1
XP_016868979.1
30
5296
XM_017013491.1
XP_016868980.1
27
4879
XM_017013492.1
XP_016868981.1
27
4876
XM_017013493.1
XP_016868982.1
29
5179
XM_017013494.1
XP_016868983.1
27
4876
XM_017013495.1
XP_016868984.1
29
5016
XM_017013496.1
XP_016868985.1
26
4599
XM_017013497.1
XP_016868986.1
27
4876
XM_017013498.1
XP_016868987.1
27
4879
XM_017013499.1
XP_016868988.1
29
5019
XM_017013500.1
XP_016868989.1
27
4876
XM_017013501.1
XP_016868990.1
29
5178
XM_017013502.1
XP_016868991.1
26
4599
GenBank mRNA
AK091406.1
-
5
653
AK295856.1
-
6
978
AK301727.1
-
5
484
AK302378.1
-
7
765
AX785390.1
-
26
4546
BC133052.1
-
26
4456
8381608
BC140946.1
-
28
4797
9056758
BQ879089.1
-
5
643
L27841.1
-
28
4955
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
neuron migration
interkinetic nuclear migration
cytoplasmic microtubule organization
microtubule anchoring
microtubule anchoring at centrosome
nonmotile primary cilium assembly
intraciliary transport involved in cilium morphogenesis
cilium assembly
negative regulation of neurogenesis
centrosome organization
protein localization to centrosome
positive regulation of intracellular protein transport
neuronal stem cell population maintenance
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-9jwpf:80/100.66.76.134:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0