Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCM1 SIRNA Assays ›
Gene Symbol:
PCM1
Gene Name:
pericentriolar material 1
Gene Aliases:
PTC4, RET/PCM-1
Chromosome Location:
Chr.8: 17922763 - 18029948 on Build GRCh38
UniGene ID:
Hs.491148
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s10129
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001315507.1
NP_001302436.1
23
4216
NM_001315508.1
NP_001302437.1
23
4219
NM_006197.3
NP_006188.3
23
4216
XM_006716336.3
XP_006716399.1
25
4598
XM_006716340.3
XP_006716403.1
25
4598
XM_006716349.3
XP_006716412.1
23
4292
XM_011544528.2
XP_011542830.1
25
4598
XM_011544529.2
XP_011542831.1
27
4904
XM_011544530.2
XP_011542832.1
26
4720
XM_011544532.2
XP_011542834.1
25
4595
XM_011544533.2
XP_011542835.1
25
4598
XM_011544534.2
XP_011542836.1
25
4598
XM_011544535.2
XP_011542837.1
25
4598
XM_011544540.2
XP_011542842.1
25
4526
XM_017013472.1
XP_016868961.1
26
4714
XM_017013473.1
XP_016868962.1
26
4710
XM_017013474.1
XP_016868963.1
24
4406
XM_017013475.1
XP_016868964.1
26
4542
XM_017013476.1
XP_016868965.1
23
4126
XM_017013477.1
XP_016868966.1
26
4710
XM_017013478.1
XP_016868967.1
26
4709
XM_017013479.1
XP_016868968.1
25
4596
XM_017013480.1
XP_016868969.1
23
4289
XM_017013481.1
XP_016868970.1
25
4592
XM_017013482.1
XP_016868971.1
23
4310
XM_017013483.1
XP_016868972.1
25
4591
XM_017013484.1
XP_016868973.1
22
4017
XM_017013485.1
XP_016868974.1
25
4595
XM_017013486.1
XP_016868975.1
24
4406
XM_017013487.1
XP_016868976.1
24
4409
XM_017013488.1
XP_016868977.1
24
4406
XM_017013489.1
XP_016868978.1
26
4709
XM_017013490.1
XP_016868979.1
26
4709
XM_017013491.1
XP_016868980.1
23
4292
XM_017013492.1
XP_016868981.1
23
4289
XM_017013493.1
XP_016868982.1
25
4592
XM_017013494.1
XP_016868983.1
23
4289
XM_017013495.1
XP_016868984.1
25
4429
XM_017013496.1
XP_016868985.1
22
4012
XM_017013497.1
XP_016868986.1
23
4289
XM_017013498.1
XP_016868987.1
23
4292
XM_017013499.1
XP_016868988.1
25
4432
XM_017013500.1
XP_016868989.1
23
4289
XM_017013501.1
XP_016868990.1
25
4591
XM_017013502.1
XP_016868991.1
22
4012
XM_017013503.1
XP_016868992.1
24
4406
XM_017013504.1
XP_016868993.1
23
4310
XM_017013505.1
XP_016868994.1
25
4429
XM_017013506.1
XP_016868995.1
23
4289
XM_017013507.1
XP_016868996.1
25
4593
XM_017013508.1
XP_016868997.1
24
4233
GenBank mRNA
AK091406.1
-
1
66
AK295856.1
-
2
391
AX785390.1
-
21
3794
BC133052.1
-
22
3869
8381608
BC140946.1
-
23
4045
9056758
BU166050.1
-
2
307
L27841.1
-
23
4203
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
neuron migration
interkinetic nuclear migration
cytoplasmic microtubule organization
microtubule anchoring
microtubule anchoring at centrosome
nonmotile primary cilium assembly
intraciliary transport involved in cilium morphogenesis
cilium assembly
negative regulation of neurogenesis
centrosome organization
protein localization to centrosome
positive regulation of intracellular protein transport
neuronal stem cell population maintenance
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-gbx7x:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0