Search
Search

Arhgdia| Gene Symbol: Arhgdia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133796.7 | NP_598557.3 | 3 | 349 | ||
| GenBank mRNA | AK075656.1 | - | 2 | 216 | ||
| AK077343.1 | - | 3 | 509 | |||
| AK143516.1 | - | 3 | 333 | |||
| AK147842.1 | - | 308 | ||||
| AK156819.1 | - | 294 | ||||
| AK165241.1 | - | 3 | 331 | |||
| AK170553.1 | - | 3 | 316 | |||
| AK170718.1 | - | 3 | 333 | |||
| AK172140.1 | - | 294 | ||||
| AK204953.1 | - | 1 | 31 | |||
| BC004732.1 | - | 3 | 291 | 3583026 | ||
| BC086755.1 | - | 3 | 273 | 6414768 | ||