Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002596.3 | NP_002587.2 | 2 | 408 | ||
NM_212502.2 | NP_997667.1 | 2 | 326 | |||
NM_212503.2 | NP_997668.1 | 2 | 326 | |||
XM_011509602.2 | XP_011507904.1 | 2 | 466 | |||
XM_017001423.1 | XP_016856912.1 | 2 | 471 | |||
GenBank mRNA | AB209307.1 | - | 1 | 803 | ||
AK293756.1 | - | 2 | 505 | |||
AK295654.1 | - | 2 | 286 | |||
AK296320.1 | - | 2 | 284 | |||
AU130437.1 | - | 2 | 287 | |||
AY353237.1 | - | 2 | 241 | |||
AY353238.1 | - | 2 | 215 | |||
BC011526.1 | - | 2 | 261 | 3855304 | ||
BM772779.1 | - | 2 | 365 | |||
BT007299.1 | - | 1 | 58 | |||
BU165355.1 | - | 328 | ||||
DC381358.1 | - | 2 | 329 |