Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bzrap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172449.2 | NP_766037.2 | 26 | 6047 | ||
GenBank mRNA | AK051895.1 | - | 5 | 1996 | ||
AK122329.1 | - | 4 | 918 | |||
BC055691.1 | - | 6 | 798 | |||
BC066146.1 | - | 5 | 1058 | 30470084 |