Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290383.1 | NP_001277312.1 | 10 | 1525 | ||
NM_194055.3 | NP_918944.2 | 10 | 1528 | |||
GenBank mRNA | AK042164.1 | - | 10 | 1140 | ||
AK140848.1 | - | 10 | 1526 | |||
BC031468.1 | - | 8 | 831 | 4037840 | ||
BC059280.1 | - | 8 | 829 | |||
DV654302.1 | - | 5 | 589 |