Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081006.1 | NP_001074475.1 | 4 | 624 | ||
NM_001177630.2 | NP_001171101.2 | 3 | 722 | |||
NM_001177631.1 | NP_001171102.1 | 3 | 722 | |||
NM_029895.4 | NP_084171.2 | 4 | 935 | |||
NM_178059.5 | NP_835160.2 | 4 | 935 | |||
GenBank mRNA | AB093289.2 | - | 3 | 512 | ||
AB125594.2 | - | 4 | 935 | |||
AB125595.2 | - | 4 | 935 | |||
AK052020.1 | - | 5 | 1037 | |||
AK052733.1 | - | 3 | 410 | |||
AK147510.1 | - | 3 | 629 | |||
BC050016.1 | - | 3 | 425 | |||
BC158051.1 | - | 3 | 444 | 9007405 | ||
BC172143.1 | - | 3 | 444 | |||
CF744489.1 | - | 4 | 624 |