Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE4D SIRNA Assays ›
Gene Symbol:
PDE4D
Gene Name:
phosphodiesterase 4D
Gene Aliases:
ACRDYS2, DPDE3, HSPDE4D, PDE43, PDE4DN2, STRK1
Chromosome Location:
Chr.5: 58969038 - 60488098 on Build GRCh38
UniGene ID:
Hs.117545
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s10200
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
7,898.00
Your Price
Online offer:
7898.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE4D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001104631.1
NP_001098101.1
7
1165
NM_001165899.1
NP_001159371.1
9
1056
NM_001197218.1
NP_001184147.1
7
904
NM_001197219.1
NP_001184148.1
7
682
NM_001197220.1
NP_001184149.1
7
1320
NM_001197221.1
NP_001184150.1
3
640
NM_001197222.1
NP_001184151.1
2
726
NM_001197223.1
NP_001184152.1
2
516
NM_006203.4
NP_006194.2
7
708
XM_005248537.3
XP_005248594.1
7
1862
XM_005248538.4
XP_005248595.1
8
3763
XM_011543469.1
XP_011541771.1
10
1145
XM_011543470.2
XP_011541772.1
10
1069
XM_011543471.1
XP_011541773.1
9
1022
XM_011543473.1
XP_011541775.1
9
970
XM_011543477.2
XP_011541779.1
9
972
XM_017009565.1
XP_016865054.1
10
1182
XM_017009566.1
XP_016865055.1
10
1072
XM_017009567.1
XP_016865056.1
8
810
XM_017009568.1
XP_016865057.1
10
1056
XM_017009569.1
XP_016865058.1
9
935
XM_017009570.1
XP_016865059.1
6
1755
XM_017009571.1
XP_016865060.1
9
978
GenBank mRNA
AF012073.1
-
7
918
AF012074.1
-
3
654
AF536975.1
-
2
323
AF536976.1
-
9
879
AF536977.1
-
7
682
AJ250852.1
-
3
250
AJ250854.1
-
8
845
AY245866.1
-
9
1021
AY245867.1
-
7
1320
AY388960.1
-
7
603
BC036319.1
-
2
518
4828512
L20970.1
-
7
716
U02882.1
-
6
486
U50157.1
-
2
324
U50158.1
-
3
238
U50159.1
-
7
659
U79571.1
-
2
729
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00174810_m1
Panther Classification:
Molecular Function -
phosphodiesterase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of heart rate
cAMP catabolic process
smooth muscle contraction
aging
regulation of receptor activity
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
cAMP-mediated signaling
neutrophil chemotaxis
regulation of cAMP metabolic process
positive regulation of interferon-gamma production
positive regulation of interleukin-2 production
positive regulation of interleukin-5 production
negative regulation of peptidyl-serine phosphorylation
multicellular organism growth
negative regulation of heart contraction
T cell receptor signaling pathway
regulation of ryanodine-sensitive calcium-release channel activity
establishment of endothelial barrier
cellular response to lipopolysaccharide
cellular response to epinephrine stimulus
adrenergic receptor signaling pathway
regulation of cardiac muscle cell contraction
adrenergic receptor signaling pathway involved in positive regulation of heart rate
regulation of cell communication by electrical coupling involved in cardiac conduction
negative regulation of relaxation of cardiac muscle
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
protein binding
drug binding
enzyme binding
cAMP binding
beta-2 adrenergic receptor binding
ion channel binding
metal ion binding
ATPase binding
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-p8fr9:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0