Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACNAT2 SIRNA Assays ›
          Gene Symbol:
          Acnat2
          Gene Name:
          acyl-coenzyme A amino acid N-acyltransferase 2
          Gene Aliases:
          C730036D15Rik
          Chromosome Location:
          Chr.4: 49375341 - 49408151 on Build GRCm38
          UniGene ID:
          Mm.35071
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s102027

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Acnat2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_145368.2 NP_663343.2 5 1136
          GenBank mRNA AK050308.1 - 5 1136
          AK050357.1 - 6 1133
          BC010829.1 - 3 980 4220620
          EU564338.1 - 3 1009

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          esterase hydrolase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          fatty acid metabolic process
          acyl-CoA metabolic process
          biological_process
          molecular_function
          dihydrolipoamide branched chain acyltransferase activity
          sterol O-acyltransferase activity
          N-acetyltransferase activity
          O-acyltransferase activity
          palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
          carnitine O-acyltransferase activity
          acetyltransferase activity
          C-acyltransferase activity
          palmitoyltransferase activity
          N-acyltransferase activity
          acylglycerol O-acyltransferase activity
          serine O-acyltransferase activity
          O-acetyltransferase activity
          O-octanoyltransferase activity
          octanoyltransferase activity
          O-palmitoyltransferase activity
          S-acyltransferase activity
          S-acetyltransferase activity
          S-malonyltransferase activity
          malonyltransferase activity
          C-palmitoyltransferase activity
          transferase activity
          transferase activity, transferring acyl groups
          succinyltransferase activity
          N-succinyltransferase activity
          O-succinyltransferase activity
          S-succinyltransferase activity
          sinapoyltransferase activity
          O-sinapoyltransferase activity
          thiolester hydrolase activity
          peptidyl-lysine N6-myristoyltransferase activity
          peptidyl-lysine N6-palmitoyltransferase activity
          benzoyl acetate-CoA thiolase activity
          3-hydroxybutyryl-CoA thiolase activity
          3-ketopimelyl-CoA thiolase activity
          N-palmitoyltransferase activity
          myristoyltransferase activity
          acyl-CoA N-acyltransferase activity
          protein-cysteine S-myristoyltransferase activity
          protein-cysteine S-acyltransferase activity
          dihydrolipoamide S-acyltransferase activity
          glucosaminyl-phosphotidylinositol O-acyltransferase activity
          ergosterol O-acyltransferase activity
          lanosterol O-acyltransferase activity
          naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
          2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
          2-methylhexanoyl-CoA C-acetyltransferase activity
          butyryl-CoA 2-C-propionyltransferase activity
          2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
          L-2-aminoadipate N-acetyltransferase activity
          UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
          keto acid formate lyase activity
          Ras palmitoyltransferase activity
          azetidine-2-carboxylic acid acetyltransferase activity
          peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
          acetyl-CoA:L-lysine N6-acetyltransferase

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-pwjgx:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0