Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177874.1 | NP_001171345.1 | 2 | 133 | ||
NM_145370.2 | NP_663345.2 | 2 | 273 | |||
GenBank mRNA | AK088490.1 | - | 2 | 130 | ||
AK190400.1 | - | 1 | 16 | |||
AK204171.1 | - | 1 | 17 | |||
BC003350.1 | - | 2 | 273 | 3481979 | ||
BC103782.1 | - | 2 | 100 | 6486599 |