Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADCY3 SIRNA Assays ›
Gene Symbol:
ADCY3
Gene Name:
adenylate cyclase 3
Gene Aliases:
AC-III, AC3
Chromosome Location:
Chr.2: 24819169 - 24920017 on Build GRCh38
UniGene ID:
Hs.467898
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s1023
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADCY3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320613.1
NP_001307542.1
10, 11
2336
NM_004036.4
NP_004027.2
10, 11
2336
XM_006711925.1
XP_006711988.1
11, 12
2118
XM_011532489.1
XP_011530791.1
11, 12
2175
XM_011532490.1
XP_011530792.1
11, 12
2175
XM_011532491.1
XP_011530793.1
10, 11
2109
XM_011532492.2
XP_011530794.1
11, 12
2175
XM_011532493.1
XP_011530795.1
11, 12
2175
XM_011532494.1
XP_011530796.1
11, 12
2118
XM_011532496.1
XP_011530798.1
10, 11
1342
XM_017003186.1
XP_016858675.1
11, 12
2118
XM_017003187.1
XP_016858676.1
10, 11
2109
XM_017003188.1
XP_016858677.1
11, 12
2175
XM_017003189.1
XP_016858678.1
11, 12
2175
XM_017003190.1
XP_016858679.1
10, 11
2052
XM_017003191.1
XP_016858680.1
10, 11
1383
XM_017003192.1
XP_016858681.1
9, 10
1219
XM_017003193.1
XP_016858682.1
9, 10
1219
GenBank mRNA
AB011083.1
-
9, 10
1172
AF033861.1
-
9, 10
1951
AK027857.1
-
9, 10
1847
AK095173.1
-
7, 8
883
AK122926.1
-
10, 11
1934
BC126235.1
-
9, 10
1862
8991951
BC144119.1
-
9, 10
1862
9052639
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00269618_m1
Gene Ontology Categories:
Function(s)
Process(es)
renal water homeostasis
cAMP biosynthetic process
female meiotic division
signal transduction
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
single fertilization
acrosome reaction
sensory perception of smell
olfactory learning
cAMP-mediated signaling
sperm motility
activation of protein kinase A activity
cellular response to glucagon stimulus
cellular response to forskolin
adenylate cyclase activity
calmodulin binding
ATP binding
calcium- and calmodulin-responsive adenylate cyclase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0