siRNA Details
Gene Symbol: Mkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177595.4 | NP_808263.2 | 3, 4 | 594 | ||
GenBank mRNA | AK034675.1 | - | 3, 4 | 572 | ||
BC137728.1 | - | 3, 4 | 529 | 8860747 |

More Information
Other Species Targeted

Panther Classification:
