Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDGFB SIRNA Assays ›
          Gene Symbol:
          PDGFB
          Gene Name:
          platelet derived growth factor subunit B
          Gene Aliases:
          IBGC5, PDGF-2, PDGF2, SIS, SSV, c-sis
          Chromosome Location:
          Chr.22: 39223359 - 39245055 on Build GRCh38
          UniGene ID:
          Hs.1976
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s10232

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PDGFB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002608.3 NP_002599.1 6 1761
          NM_033016.3 NP_148937.1 6 661
          GenBank mRNA BC029822.1 - 6 981 5174750
          BC077725.1 - 6 1493 6379783
          CR456538.1 - 6 677
          CR541807.1 - 6 669
          M12783.1 - 6 1761
          X02744.1 - 6 786
          X02811.1 - 6 1651
          X03702.1 - 5 594

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          response to hypoxia
          embryonic placenta development
          positive regulation of endothelial cell proliferation
          monocyte chemotaxis
          platelet degranulation
          positive regulation of glomerular filtration
          DNA replication
          protein phosphorylation
          transforming growth factor beta receptor signaling pathway
          heart development
          positive regulation of cell proliferation
          response to wounding
          negative regulation of phosphatidylinositol biosynthetic process
          negative regulation of platelet activation
          positive regulation of gene expression
          negative regulation of gene expression
          regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of smooth muscle cell migration
          cell growth
          peptidyl-serine phosphorylation
          peptidyl-tyrosine phosphorylation
          hemopoiesis
          extracellular matrix organization
          positive regulation of cell migration
          positive regulation of protein autophosphorylation
          negative regulation of protein binding
          activation of protein kinase activity
          activation of protein kinase B activity
          response to estradiol
          response to insulin
          positive regulation of metanephric mesenchymal cell migration by platelet-derived growth factor receptor-beta signaling pathway
          paracrine signaling
          wound healing
          response to drug
          positive regulation of MAP kinase activity
          positive regulation of MAPK cascade
          positive regulation of blood vessel endothelial cell migration
          positive regulation of GTPase activity
          positive regulation of phosphatidylinositol 3-kinase activity
          response to estrogen
          positive regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of DNA replication
          positive regulation of mitotic nuclear division
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          phosphatidylinositol phosphorylation
          platelet-derived growth factor receptor signaling pathway
          phosphatidylinositol-mediated signaling
          positive regulation of fibroblast proliferation
          positive regulation of smooth muscle cell proliferation
          positive regulation of peptidyl-tyrosine phosphorylation
          positive chemotaxis
          positive regulation of chemotaxis
          positive regulation of cell division
          cell chemotaxis
          positive regulation of protein tyrosine kinase activity
          positive regulation of ERK1 and ERK2 cascade
          protein kinase C signaling
          cellular response to growth factor stimulus
          cellular response to mycophenolic acid
          positive regulation of glomerular mesangial cell proliferation
          metanephric glomerular mesangial cell development
          reactive oxygen species metabolic process
          positive regulation of calcium ion import
          positive regulation of hyaluronan biosynthetic process
          negative regulation of pri-miRNA transcription from RNA polymerase II promoter
          positive regulation of pri-miRNA transcription from RNA polymerase II promoter
          positive regulation of vascular smooth muscle cell proliferation
          positive regulation of vascular associated smooth muscle cell migration
          negative regulation of vascular smooth muscle cell differentiation
          positive regulation of vascular smooth muscle cell dedifferentiation
          positive regulation of reactive oxygen species metabolic process
          positive regulation of DNA biosynthetic process
          positive regulation of metanephric mesenchymal cell migration
          Ras guanyl-nucleotide exchange factor activity
          platelet-derived growth factor receptor binding
          protein binding
          collagen binding
          growth factor activity
          superoxide-generating NADPH oxidase activator activity
          chemoattractant activity
          identical protein binding
          protein homodimerization activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity
          protein heterodimerization activity
          platelet-derived growth factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0