Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145962.2 | NP_666074.1 | 4 | 1161 | ||
GenBank mRNA | AK039865.1 | - | 4 | 1163 | ||
AK136305.1 | - | 4 | 851 | |||
AK163377.1 | - | 3 | 548 | |||
BC027089.1 | - | 4 | 773 | 5355443 | ||
BC032188.1 | - | 4 | 1207 |