Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADCY5 SIRNA Assays ›
          Gene Symbol:
          ADCY5
          Gene Name:
          adenylate cyclase 5
          Gene Aliases:
          AC5, FDFM
          Chromosome Location:
          Chr.3: 123282296 - 123448632 on Build GRCh38
          UniGene ID:
          Hs.593292
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1025

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ADCY5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199642.1 NP_001186571.1 7 858
          NM_183357.2 NP_899200.1 7 1833
          XM_005247077.3 XP_005247134.1 7 1920
          XM_005247078.1 XP_005247135.1 7 858
          XM_006713483.1 XP_006713546.1 7 854
          XM_006713484.1 XP_006713547.1 7 859
          XM_011512359.2 XP_011510661.1 7 1718
          XM_011512360.2 XP_011510662.1 7 915
          XM_011512361.1 XP_011510663.1 7 822
          XM_017005638.1 XP_016861127.1 7 750
          XM_017005639.1 XP_016861128.1 7 750
          XM_017005640.1 XP_016861129.1 7 846
          GenBank mRNA AF497517.1 - 7 734
          AK093840.1 - 8 822
          AK124691.1 - 8 1270
          AK294565.1 - 7 1026
          AK303070.1 - 7 859
          BX647739.1 - 4 335
          U65473.1 - 2 157

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          adenosine receptor signaling pathway
          renal water homeostasis
          cAMP biosynthetic process
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          activation of adenylate cyclase activity
          adenylate cyclase-activating dopamine receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          adenylate cyclase-inhibiting dopamine receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          locomotory behavior
          cAMP-mediated signaling
          activation of protein kinase A activity
          neuromuscular process controlling balance
          regulation of insulin secretion involved in cellular response to glucose stimulus
          cellular response to glucagon stimulus
          cellular response to forskolin
          adenylate cyclase activity
          ATP binding
          adenylate cyclase binding
          metal ion binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0