Search
Search

Vstm2a| Gene Symbol: Vstm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290539.1 | NP_001277468.1 | 3, 4 | 579 | ||
| NM_145967.2 | NP_666079.2 | 3, 4 | 579 | |||
| GenBank mRNA | AK032473.1 | - | 3, 4 | 578 | ||
| AK033048.1 | - | 3, 4 | 581 | |||
| AK079096.1 | - | 501 | ||||
| AK137363.1 | - | 3, 4 | 501 | |||
| BC027127.1 | - | 3, 4 | 531 | 5361611 | ||
| BM963943.1 | - | 3, 4 | 239 | |||