Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ENPP2 SIRNA Assays ›
          Gene Symbol:
          ENPP2
          Gene Name:
          ectonucleotide pyrophosphatase/phosphodiesterase 2
          Gene Aliases:
          ATX, ATX-X, AUTOTAXIN, LysoPLD, NPP2, PD-IALPHA, PDNP2
          Chromosome Location:
          Chr.8: 119557077 - 119673404 on Build GRCh38
          UniGene ID:
          Hs.190977
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s10267

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ENPP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001040092.2 NP_001035181.1 24 2387
          NM_001130863.2 NP_001124335.1 25 2462
          NM_006209.4 NP_006200.3 25 2543
          NR_045555.1 13 1292
          XM_006716584.1 XP_006716647.1 26 2596
          XM_006716585.1 XP_006716648.1 26 2584
          XM_006716587.1 XP_006716650.1 25 2509
          XM_017013571.1 XP_016869060.1 25 2496
          XM_017013572.1 XP_016869061.1 25 2428
          XM_017013573.1 XP_016869062.1 25 2484
          XM_017013574.1 XP_016869063.1 24 2421
          XM_017013575.1 XP_016869064.1 24 2353
          GenBank mRNA AK124910.1 - 6 2922
          AK130313.1 - 13 1292
          AK296027.1 - 10 1003
          AK315253.1 - 24 2360
          AL544867.3 - 7 689
          BC034961.2 - 24 2360 4828484
          EU131011.2 - 25 2376
          L35594.1 - 25 2506

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          phosphate-containing compound metabolic process
          receptor-mediated endocytosis
          movement of cell or subcellular component
          chemotaxis
          immune response
          G-protein coupled receptor signaling pathway
          phospholipid catabolic process
          positive regulation of epithelial cell migration
          regulation of cell migration
          phosphatidylcholine catabolic process
          regulation of angiogenesis
          positive regulation of peptidyl-tyrosine phosphorylation
          nucleic acid phosphodiester bond hydrolysis
          positive regulation of lamellipodium morphogenesis
          nucleic acid binding
          phosphodiesterase I activity
          nucleotide diphosphatase activity
          lysophospholipase activity
          scavenger receptor activity
          calcium ion binding
          transcription factor binding
          zinc ion binding
          hydrolase activity
          polysaccharide binding
          alkylglycerophosphoethanolamine phosphodiesterase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-hlnc5:80/100.66.79.239:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0