Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDZK1 SIRNA Assays ›
          Gene Symbol:
          PDZK1 PDZK1P1
          Gene Name:
          PDZ domain containing 1
          PDZ domain containing 1 pseudogene 1
          Gene Aliases:
          CAP70, CLAMP, NHERF-3, NHERF3, PDZD1
          OTTHUMT00000038524, PDZK1P2
          Chromosome Location:
          Chr.1: 145670852 - 145707507 on Build GRCh38
          Chromosome Location:
          Chr.1: 147993862 - 148014956 on Build GRCh38
          UniGene ID:
          Hs.444751
          Hs.696575
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s10284

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PDZK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001201325.1 NP_001188254.1 3 424
          NM_001201326.1 NP_001188255.1 3 424
          NM_002614.4 NP_002605.2 4 495
          XM_011509616.2 XP_011507918.1 2 340
          XM_011509618.2 XP_011507920.1 3 428
          XM_011509619.2 XP_011507921.2 2 340
          XM_017001464.1 XP_016856953.1 2 340
          XM_017001465.1 XP_016856954.1 2 340
          XM_017001466.1 XP_016856955.1 2 340
          XM_017001467.1 XP_016856956.1 3 531
          GenBank mRNA AF012281.1 - 3 371
          AK298269.1 - 3 365
          AK313100.1 - 3 376
          BC006496.2 - 3 435 2959384
          BC006518.2 - 3 435 2959384
          BX477678.1 - 4 494
          Gene Symbol: PDZK1P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_111936.1 4 533
          GenBank mRNA AK126719.1 - 3 1902
          DA085292.1 - 4 533

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          cell proliferation
          carnitine transport
          drug transport
          positive regulation of ion transmembrane transport
          regulation of anion transport
          positive regulation of protein targeting to membrane
          scavenger receptor binding
          transporter activity
          protein binding
          PDZ domain binding
          protein complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-6qvzt:80/100.66.73.36:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0