Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297612.1 | NP_001284541.1 | 4, 5 | 1088 | ||
NM_175441.5 | NP_780650.2 | 4, 5 | 1541 | |||
GenBank mRNA | AK052858.1 | - | 4, 5 | 1376 | ||
AK146683.1 | - | 4, 5 | 1542 | |||
AK165216.1 | - | 4, 5 | 1030 | |||
BC137983.1 | - | 4, 5 | 1514 | 8861002 | ||
BC145079.1 | - | 4, 5 | 1515 | |||
EU403565.1 | - | 4, 5 | 1484 |