Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297612.1 | NP_001284541.1 | 9 | 1573 | ||
NM_175441.5 | NP_780650.2 | 9 | 2026 | |||
GenBank mRNA | AK031546.1 | - | 2 | 66 | ||
AK052858.1 | - | 9 | 1861 | |||
AK146683.1 | - | 9 | 2027 | |||
AK165216.1 | - | 9 | 1515 | |||
BC137983.1 | - | 9 | 1999 | 8861002 | ||
BC145079.1 | - | 9 | 2000 | |||
EU403565.1 | - | 9 | 1969 |