Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000288.3 | NP_000279.1 | 6 | 672 | ||
XM_005267019.4 | XP_005267076.1 | 6 | 952 | |||
GenBank mRNA | BC006268.1 | - | 6 | 614 | 3948721 | |
BC031606.1 | - | 6 | 656 | 5176358 | ||
CR542054.1 | - | 6 | 570 | |||
CR542076.1 | - | 6 | 570 | |||
U76560.1 | - | 6 | 618 | |||
U88871.1 | - | 6 | 651 |