Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145413.4 | NP_663388.1 | 4, 5 | 752 | ||
GenBank mRNA | AK049705.1 | - | 3, 4 | 554 | ||
AK129154.1 | - | 2, 3 | 161 | |||
AK132149.1 | - | 4, 5 | 753 | |||
BC019381.1 | - | 4, 5 | 771 | 4206622 | ||
BC023737.1 | - | 5, 6 | 968 | 5352660 | ||
BC024412.1 | - | 3, 4 | 702 | 4971483 | ||
BC031473.1 | - | 4, 5 | 864 | 3990487 |