Search
Search

Cnksr3| Gene Symbol: Cnksr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172546.2 | NP_766134.1 | 13 | 2073 | ||
| GenBank mRNA | AK033015.1 | - | 13 | 2074 | ||
| AK149951.1 | - | 12 | 1372 | |||
| AY151137.1 | - | 13 | 2035 | |||
| BC024086.1 | - | 12 | 1421 | |||
| BC031194.1 | - | 6 | 1180 | 5059479 | ||