Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033259.4 | NP_001028431.2 | 7, 8 | 990 | ||
GenBank mRNA | AK161322.1 | - | 4, 5 | 812 | ||
BC139254.1 | - | 4, 5 | 607 | 8862276 | ||
BC139257.1 | - | 4, 5 | 607 | 8862279 | ||
DV649183.1 | - | 2, 3 | 261 |