Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbno2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183426.1 | NP_906271.1 | 7 | 778 | ||
GenBank mRNA | AJ512611.1 | - | 8 | 934 | ||
AK089496.1 | - | 4 | 523 | |||
AK149806.1 | - | 7 | 802 | |||
AK155462.1 | - | 7 | 776 | |||
AK155674.1 | - | 7 | 775 | |||
AK170733.1 | - | 6 | 570 | |||
BC056369.1 | - | 7 | 778 | 6831600 | ||
BC064113.1 | - | 4 | 535 |