siRNA Details
Gene Symbol: Gabrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146017.3 | NP_666129.1 | 10 | 1405 | ||
GenBank mRNA | AK028946.1 | - | 10 | 1405 | ||
AK160577.1 | - | 10 | 1409 | |||
BC023693.1 | - | 10 | 1392 | 5344479 | ||
BC025550.1 | - | 10 | 1350 | 5320737 | ||
BC027245.1 | - | 10 | 1368 | 3602400 | ||
BC027839.1 | - | 6 | 957 | |||
BC031196.1 | - | 10 | 1379 | 5059564 |

More Information
Panther Classification:
Molecular Function -






