Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153788.3 | NP_722483.2 | 17 | 1865 | ||
GenBank mRNA | AK029819.1 | - | 5 | 2639 | ||
AK133473.1 | - | 12 | 3063 | |||
AK138736.1 | - | 17 | 1858 | |||
AK154449.1 | - | 17 | 1868 | |||
AK155001.1 | - | 17 | 1886 | |||
BC031462.1 | - | 17 | 1865 | 4017986 | ||
BC037481.1 | - | 12 | 1492 | 4460639 | ||
BC064116.1 | - | 3 | 1148 |