Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167976.1 | NP_001161448.1 | 7 | 466 | ||
NM_172577.3 | NP_766165.2 | 6 | 909 | |||
GenBank mRNA | AK036988.1 | - | 6 | 649 | ||
AK041694.1 | - | 6 | 910 | |||
AK045180.1 | - | 6 | 735 | |||
AK162379.1 | - | 6 | 736 | |||
AK167535.1 | - | 6 | 617 | |||
BC057980.1 | - | 6 | 733 | 5042653 | ||
EU180236.1 | - | 7 | 466 |