Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mis18bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172578.2 | NP_766166.2 | 7 | 1637 | ||
GenBank mRNA | AK032698.1 | - | 5 | 668 | ||
AK035117.1 | - | 7 | 1636 | |||
AK220223.1 | - | 6 | 903 | |||
BC021832.1 | - | 1 | 180 | |||
BC052175.1 | - | 7 | 1637 | 6394068 |