siRNA Details
Gene Symbol: Sox21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177753.3 | NP_808421.1 | 1 | 2846 | ||
GenBank mRNA | AY069926.1 | - | 1 | 1665 | ||
AY142959.1 | - | 1 | 1718 | |||
BC063054.1 | - | 2 | 1634 | |||
BC069176.1 | - | 2 | 2147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
