Search
Search

Selo| Gene Symbol: Selo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027905.2 | NP_082181.2 | 2 | 681 | ||
| GenBank mRNA | AK005048.1 | - | 2 | 681 | ||
| AK054432.1 | - | 2 | 701 | |||
| AK088096.1 | - | 2 | 706 | |||
| AK161059.1 | - | 2 | 690 | |||
| BC117817.1 | - | 3 | 196 | |||
| BC117818.1 | - | 3 | 229 | |||
| BC141293.1 | - | 2 | 750 | 9055905 | ||