Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poglut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300827.1 | NP_001287756.1 | 8, 9 | 1088 | ||
NM_172380.4 | NP_759012.1 | 8, 9 | 885 | |||
GenBank mRNA | AK031608.1 | - | 8, 9 | 824 | ||
AK035948.1 | - | 8, 9 | 833 | |||
AK036224.1 | - | 8, 9 | 815 | |||
AK041321.1 | - | 8, 9 | 823 | |||
AK045278.1 | - | 8, 9 | 1033 | |||
AK079886.1 | - | 8, 9 | 855 | |||
BC026809.1 | - | 8, 9 | 836 | 4205469 |