Search Thermo Fisher Scientific
Gene Symbol: Npas4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153553.4 | NP_705781.1 | 6 | 1047 | ||
GenBank mRNA | AB049835.1 | - | 6 | 942 | ||
AK032607.1 | - | 4 | 1561 | |||
AY730724.1 | - | 6 | 1074 | |||
BC129861.1 | - | 6 | 983 | 40110003 |