Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PIK3R1 SIRNA Assays ›
          Gene Symbol:
          PIK3R1
          Gene Name:
          phosphoinositide-3-kinase regulatory subunit 1
          Gene Aliases:
          AGM7, GRB1, IMD36, p85, p85-ALPHA
          Chromosome Location:
          Chr.5: 68215737 - 68301821 on Build GRCh38
          UniGene ID:
          Hs.132225
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s10536
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PIK3R1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_181504.3 NP_852556.2 2 245
          NM_181523.2 NP_852664.1 8 1545
          NM_181524.1 NP_852665.1 2 377
          XM_005248542.3 XP_005248599.1 8 1568
          XM_011543493.2 XP_011541795.1 7 1198
          XM_017009585.1 XP_016865074.1 8 2337
          XM_017009586.1 XP_016865075.1 7 1158
          GenBank mRNA AF279367.1 - 2 377
          AK000121.1 - 2 291
          AK126345.1 - 1 1508
          AK223613.1 - 2 225
          AK299743.1 - 6 574
          BC030815.1 - 2 217 4290954
          BC094795.1 - 8 1525 30528412
          DA487709.1 - 2 245
          M61906.1 - 7 1007

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00381459_m1

          Panther Classification:

          Molecular Function -

          kinase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular glucose homeostasis
          negative regulation of cell-matrix adhesion
          protein phosphorylation
          phosphatidylinositol biosynthetic process
          epidermal growth factor receptor signaling pathway
          insulin receptor signaling pathway
          extrinsic apoptotic signaling pathway via death domain receptors
          intrinsic apoptotic signaling pathway in response to DNA damage
          phosphatidylinositol 3-kinase signaling
          regulation of phosphatidylinositol 3-kinase signaling
          viral process
          platelet activation
          B cell differentiation
          positive regulation of cell migration
          T cell costimulation
          positive regulation of tumor necrosis factor production
          cellular response to insulin stimulus
          positive regulation of RNA splicing
          cellular response to UV
          response to endoplasmic reticulum stress
          phosphatidylinositol-3-phosphate biosynthetic process
          Fc-epsilon receptor signaling pathway
          Fc-gamma receptor signaling pathway involved in phagocytosis
          ERBB2 signaling pathway
          positive regulation of transcription factor import into nucleus
          negative regulation of apoptotic process
          regulation of phosphatidylinositol 3-kinase activity
          negative regulation of osteoclast differentiation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of glucose import
          phosphatidylinositol phosphorylation
          insulin-like growth factor receptor signaling pathway
          vascular endothelial growth factor receptor signaling pathway
          phosphatidylinositol-mediated signaling
          protein stabilization
          T cell receptor signaling pathway
          leukocyte migration
          regulation of stress fiber assembly
          NFAT protein import into nucleus
          growth hormone receptor signaling pathway
          positive regulation of establishment of protein localization to plasma membrane
          positive regulation of endoplasmic reticulum unfolded protein response
          positive regulation of glucose import in response to insulin stimulus
          transmembrane receptor protein tyrosine kinase adaptor activity
          insulin receptor binding
          insulin-like growth factor receptor binding
          neurotrophin TRKA receptor binding
          protein binding
          transcription factor binding
          1-phosphatidylinositol-3-kinase activity
          protein phosphatase binding
          phosphatidylinositol 3-kinase regulator activity
          phosphatidylinositol 3-kinase regulatory subunit binding
          ErbB-3 class receptor binding
          phosphatidylinositol 3-kinase binding
          insulin binding
          insulin receptor substrate binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity
          1-phosphatidylinositol-3-kinase regulator activity
          protein heterodimerization activity

          Back To Top
           

          Citations (1)

          Nuclear degradation of Wilms tumor 1-associating protein and survivin splice variant switching underlie IGF-1-mediated survival.
          Authors
          Small TW, Pickering JG
          Journal
          J Biol Chem - Page No 24684 (Fri Sep 11 00:00:00 UTC 2009)
          Volume (Year)
          284 (2009)
          Product Usage
          -

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0